Learn More
Promega MagaZorb™ RNA Kit
Catalog No. PRMB2004 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
200 Preps
Catalog No. Quantity
PRMB2004 200 Preps
1 options
Catalog No. PRMB2004
Supplier: Promega MB2004
Includes
Proteinase K, Lysis buffer, Binding buffer, MagaZorb reagent, Wash buffer, RNase-free water
The MagaZorb RNA Kit provides an easy, fast and cost-effective technique for isolating PCR-quality total RNA.
  • Fast, Cost-Effective Isolation of PCR-Quality RNA
  • RNA from whole blood (fresh or citrate-, heparin- or EDTA-treated), buffy coat, leukocytes or tissues
  • No need for centrifugation, vacuum filtration or column separation
  • No organic solvents
Specifications
Content And Storage 15°C to 30°C
Includes Proteinase K solution 4mL, Lysis Buffer 40mL, Binding Buffer 100mL, MagaZorb™ reagent 4mL, Wash Buffer 200mL, RNase free Water 40mL
Quantity 200 Preps
U.S. Patent 6,855,499 and other patents pending.