Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega MagnaBot™ II Magnetic Separation Device

Catalog No. PRV8351 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8351 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV8351 Supplier Promega Supplier No. V8351
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The MagnaBot II Magnetic Separation Device is designed for high-throughput bioseparation using magnetic particles such as MagneSil™ Paramagnetic Particles with a 96-well PCR plate.

  • Designed to work with a 96-well PCR plate
  • Allows the quantitative removal of liquids

†Patent Pending.

Specifications

Content And Storage 15°C to 30°C
Isolation Technology Magnetic Bead
For Use With (Equipment) Magnesil™ paramagnetic particles and a PCR plate
Product Type Accessory, Magnetic Separation
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.