Learn More
Promega MagneSil™ Genomic, Fixed-Tissue System
Catalog No. PRMD1490 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Magnesil Genomic Fixed Tissue; 100 preps
Catalog No.
PRMD1490 Magnesil Genomic Fixed Tissue; 100 preps
1 options
Catalog No. PRMD1490 Supplier Promega Supplier No. MD1490
Includes
Proteinase K, DTT, and Incubation Buffer (shipped on dry ice and stored at -20°C); Lysis Buffer, 2X Wash Buffer, Resin, and Elution Buffer (shipped and stored at room temperature)
MagneSil™ Genomic, Fixed Tissue System provides a fast, simple technique to prepare genomic DNA from formalin-fixed, paraffin-embedded tissue.
  • Magnetic Method to Purify DNA from FFPE Tissue
  • One-hour purification after overnight proteinase K digestion
  • No centrifugation of lysate required
  • Amplifiable DNA from 10µM FFPE tissue slices