Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega NanoLuc™ pNL1 Vectors

Catalog No. PRN1101 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
pNL1.1(Nluc)
pNL1.1.CMV(Nluc/CMV)
pNL1.2(NlucP)
pNL1.3(secNluc)
pNL1.3.CMV(secNluc/CMV)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRN1101 pNL1.3.CMV(secNluc/CMV)
PRN1001 pNL1.1(Nluc)
PRN1011 pNL1.2(NlucP)
PRN1021 pNL1.3(secNluc)
PRN1091 pNL1.1.CMV(Nluc/CMV)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PRN1101 Supplier Promega Supplier No. N1101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Configured with NanoLuc Luciferase for use in reporter gene assays of transcriptional control—ideal for cloning of known or putative promoter region

  • Vectors for Expressing a Secreted Reporter Protein in Mammalian Cells
  • NanoLuc(R) luciferase reporter modified with a secretion sequence for extracellular expression
  • Select different promoter options for cloning a putative regulatory element or use as a control reporter vector
  • One vector offers hygromycin selection for stable expression

Specifications

Product Type pNL1.3.CMV(secNluc/CMV)
pH 8
Content And Storage -30°C to -10°C
Quantity 20 μg
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.