Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega NanoLuc™ pNL1 Vectors

Catalog No. p-4671693
Click to view available options
Product Type:
pNL1.1(Nluc)
pNL1.1.CMV(Nluc/CMV)
pNL1.2(NlucP)
pNL1.3(secNluc)
pNL1.3.CMV(secNluc/CMV)

Configured with NanoLuc Luciferase for use in reporter gene assays of transcriptional control—ideal for cloning of known or putative promoter region

  • Insert a Putative Promoter to Drive Expression of NanoLuc(R) Luciferase
  • Multiple cloning site offers an array of restriction sites for cloning
  • Offers two engineered genes for your reporter assay: Nluc and NlucP
  • NanoLuc(R) enzyme and its substrate are about 100-fold brighter than other luciferases, producing a high intensity, glow-type luminescence

Specifications

Product Type pNL1.1(Nluc)
pH 8
Content And Storage -30°C to -10°C
Quantity 20 μg
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.