Learn More
Description
- Clone a Putative Promoter for Expression in Mammalian Cells and Select Stable Transfectants
- Offers two engineered genes for your reporter assay: Nluc and NlucP
- NanoLuc(R) enzyme and its substrate are about 100-fold brighter than other luciferases, producing a high intensity, glow-type luminescence
- Use for transient transfection or select for stable expression with hygromycin
Specifications
Specifications
| Product Type | pNL2.2(NlucP/Hygro) |
| pH | 8 |
| Content And Storage | -30°C to -10°C |
| Quantity | 20 μg |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.