Learn More
Promega p70S6K Kinase Enzyme System
Catalog No. PRV2741 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV2741 10 μg
1 options
Catalog No. PRV2741 Supplier Promega Supplier No. V2741
Includes
  • Kinase: p70S6K, 10μg (Human, recombinant full-length), M.W.: ∽76kDa
  • Substrate: S6K substrate (KRRRLASLR); derived from human 40S ribosomal protein S6 (amino acid 230–238)
  • Others: Reaction Buffer, DTT
Recombinant full-length human p70S6K was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. p70S6K is responsible for the phosphorylation of 40S ribosomal protein S6 and is ubiquitously expressed in human adult tissues.
  • Easily Screen and Profile p70S6K Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg