Learn More
Promega PDGFRα Kinase Enzyme System
Catalog No. PRV8011 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ADP-Glo™ Kinase Assay, PDGFRα Kinase Enzyme System
PDGFRα Kinase Enzyme System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV8011 1/Ea. ADP-Glo™ Kinase Assay, PDGFRα Kinase Enzyme System
PRV3721 10 μg PDGFRα Kinase Enzyme System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV8011 Supplier Promega Supplier No. V8011
Includes
  • Kinase: PDGFRα, 10μg (Human, recombinant; amino acids 550–end), M.W.: ∽95kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT
Recombinant human PDGFR alpha (amino acids 550-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFR alpha is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity.
  • Easily Screen and Profile PDGFRα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Product Type ADP-Glo™ Kinase Assay, PDGFRα Kinase Enzyme System
Content And Storage Components at different temps
Quantity 1/Ea.