Learn More
Promega PDGFRβ Kinase Enzyme System
Catalog No. PRV8021 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8021 Each
PRV3731 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV8021 Supplier Promega Supplier No. V8021
Recombinant human PDGFR beta (amino acids 557-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFR beta is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity.
  • Easily Screen and Profile PDGFRβ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Includes 1 x 10μg PDGFRbeta Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay
Content And Storage Store the Kinase at -70°C and the rest of the components at -20°C
For Use With (Application) Elevated in several cancers and inhibition of PDGFRbeta activity blocks progression of renal carcinoma in an animal model
Quantity Each