Learn More
Promega PKCβ II Kinase Enzyme System
Catalog No. PRV9701 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9701 1/Ea.
PRV3741 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9701 Supplier Promega Supplier No. V9701
Recombinant full-length human PKC beta II was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC beta II is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases.
  • Easily Screen and Profile PKCβII Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Includes 1 x 10μg PKCbetaII Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay
Content And Storage Components at different temps
Quantity 1/Ea.