Learn More
Promega PKCα Kinase Enzyme System
Catalog No. PRV3381 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV3381 10 μg
1 options
Catalog No. PRV3381 Supplier Promega Supplier No. V3381
Includes
  • Kinase: PKCα, 10μg (Human, recombinant; amino acids 550–end), M.W.: ∽103kDa
  • Substrate: CREBtide (KRREILSRRPSYR); derived from human CREB1 isoform A (amino acid 109–121)
  • Others: Reaction Buffer, DTT, Lipid solution
Recombinant full-length human PKC alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC alpha is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases.
  • Easily Screen and Profile PKCα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg