Learn More
Promega ADP-Glo™ Kinase Assay + PKC alpha Kinase Enzyme System
Catalog No. PRV9691 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
PRV9691 1/Ea.
1 options
Catalog No. PRV9691 Supplier Promega Supplier No. V9691
Recombinant full-length human PKC alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC alpha is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases.
  • Easily Screen and Profile PKCα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage Components at different temps
Quantity 1/Ea.