Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ProFection™ Mammalian Transfection System

Catalog No. PRE1200 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
40 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-E1200 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-E1200 Supplier Promega Supplier No. E1200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calcium Phosphate-mediated transfection reagent used for transfection of DNA into many cell types.

  • Calcium Phosphate-Mediated Transfection Method
  • Sufficient reagents for 40 high-efficiency transfections of cells plated in 100mm tissue culture dishes
  • High-efficiency stable or transient transfection of eukaryotic cells

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) Production of long-term stable transfectants, also work well for transient expression of transfected genes and can be used with most adherent cell line
Includes Nuclease-free Water, 2M Calcium Chloride, Hepes 2X Buffered Saline
No. of Reactions 40
Quantity 40 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.