Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Promega™-Markers™ Lambda Ladder

Catalog No. PRG3011 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
40 to 60 Lanes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-G3011 40 to 60 Lanes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PR-G3011 Supplier Promega Supplier No. G3011
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

50-1,000kb; 20 bands. Concatamer of lambda phage DNA. Embedded in dye-colored, 1% agarose string molds in 50mM EDTA at 0.5ug/5mm.

  • λ Phage DNA Concatemerized into Multimers of 50-800kb+
  • Embedded in dye-colored, 0.5% agarose string molds in 50mM EDTA
  • DNA concentration is 0.5μg/5mm
  • Certified for use as markers in pulsed-field gel electrophoresis

Specifications

Concentration 0.5 μg/5 mm
Form Liquid
Formulation Embedded in dye-colored, 1% Agarose string molds in 50mM EDTA
For Use With (Application) Pulsed-field analysis of chromosomal DNA
No. of Bands 20
Quantity 40 to 60 Lanes
Storage Requirements 2 to 10°C
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.