Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Proteasome-Glo™ 3 Assay

Catalog No. PRG1180 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG1180 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PRG1180 Supplier Promega Supplier No. G1180
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Proteasome-Glo™ Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells.

  • Homogeneous, Luminescent Assays that Measure Protease Activities in Cultured Cells
  • Simple add-mix-measure protocol
  • Chymotrypsin-Like, Trypsin-Like and Caspase-Like activity data directly from cellular environment
  • Results within 10-30 minutes after reagent addition

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) Cell permeabilization, proteasome activity and Luciferase activity
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.