Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Proteasome-Glo™ Bioluminescent Enzyme Assays

Catalog No. p-4390071 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
Product Type:
3-Substrate System
Caspase-like Assay
Chymotrypsin-like Assay
Trypsin-like Assay
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRG8621 10 mL Chymotrypsin-like Assay
PRG8622 50 mL Chymotrypsin-like Assay
PRG8631 10 mL Trypsin-like Assay
PRG8641 10 mL Caspase-like Assay
PRG8531 10 mL 3-Substrate System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PRG8621 Supplier Promega Supplier No. G8621
Only null left
Add to Cart
Add to Cart

The Proteasome-Glo Assays are homogeneous, luminescent assays that individually measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome in a purified enzyme-based format (not suitable for cell-based applications).

  • Measure Chymotrypsin-, Trypsin- and Caspase-Like Activities
  • Homogeneous, luminescent enzyme-based assays
  • Simple add-mix-measure protocol
  • Results in 10-30 minutes after reagent addition

Specifications

Product Type Chymotrypsin-like Assay
Includes 1 x 10mL Proteasome Glo Chymotrypsin, 1 x 50μL Suc LLVY Glo substrate, 1 x 1 each Luciferin detection Reagent
Detection Method Bioluminescence
pH 7.2
Content And Storage -30°C to -10°C
Product Line Proteasome-Glo
Quantity 10 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.