Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ProTEV Plus Protease

Catalog No. p-4389612 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 U
8000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6101 1000 U
PRV6102 8000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV6101 Supplier Promega Supplier No. V6101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ProTEV Plus is an improved 48kDa version of the Nla protease from tobacco etch virus (TEV) that has been engineered to be more stable than native TEV protease for prolonged enzymatic activity.

  • Engineered Protease with Greater Stability, Prolonged Activity Over Native TEV Protease
  • Active over broad pH (5.5-8.5) and temperature (4-30°C) range
  • Cleaves protein fusions with the ENLYFQ(G/S) sequence
  • Use with in-solution and on-column digestions
  • Supplied at a concentration of 5u/μL

Specifications

Concentration 5 U/μL
For Use With (Application) Cleavage of affinity tags from fusion Proteins after Protein purification, removal of HaloTag™ Protein during Protein purification/expression of used Proteins of interest
Kit Contents HQ tag located at N-terminus to be immobilized on affinity resins
Content And Storage -30°C to -10°C
Quantity 1000 U
Product Type Protease
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.