Learn More
Promega pSP73 Vectors
Catalog No. PRP2221 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
pSP73 Vector
Catalog No.
PRP2221 pSP73 Vector
1 options
Catalog No. PRP2221 Supplier Promega Supplier No. P2221
Used as standard cloning vector or for transcription of RNA in vitro.
  • Standard Cloning Vector That Also Transcribes RNA In Vitro
  • Versatile vector that can be used for standard cloning and in vitro transcription
  • SP6 and T7 RNA polymerase promoters flank the multiple cloning region
  • Multiple cloning site provides a convenient selection of restriction sites for cloning