Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega pSV-β-Galactosidase Control Vector

Catalog No. PRE1081 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-E1081 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-E1081 Supplier Promega Supplier No. E1081
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A positive control vector for monitoring transfection efficiencies of mammalian cells. The SV40 early promoter and enhancer drive transcription of the lacZ gene, which encodes the beta-galactosidase enzyme.

  • Monitors Transfection Efficiencies of Mammalian Cells
  • Co-transfect into cells as the control reporter in a dual-reporter assay
  • Assay in cell extracts using spectrophotometric, fluorescent or chemiluminescent methods
  • Can be used for in situ histochemical analysis using X-gal substrate

Specifications

Promoter SV40
Reporter Gene lacZ
Content And Storage -30°C to -10°C
Quantity 20 μg
Vector pSV-beta-Galactosidase control vector
For Use With (Application) Monitoring transfection efficiencies of mammalian cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.