Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega pTNT™ Vector

Catalog No. PRL5610 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRL5610 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRL5610 Supplier Promega Supplier No. L5610
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The pCMVTNT and pTNT Vectors are designed for convenient expression of cloned genes in vitro or in vivo. SP6 and T7 promoters allow expression from SP6- or T7-based coupled in vitro transcription/translation systems.

  • Convenient Expression of Cloned Genes In Vitro or In Vivo
  • Express cloned genes in vitro or in vivo from SP6- or T7-based coupled in vitro transcription/translation systems

Specifications

Promoter SP6, T7
Content And Storage Below -10°C
Quantity 20 μg
Vector pTnT vector
For Use With (Application) Appropriate in-vitro translation or transcription system

Licensed for research and laboratory use only under U.S. Patent 5,547,862.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.