Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega QuantiFluor™ dsDNA System

Catalog No. PRE2670 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE2670 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE2670 Supplier Promega Supplier No. E2670
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantification of double-stranded DNA using fluorescent methods

The QuantiFluor™ dsDNA System contains a fluorescent DNA-binding dye that enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA) in solution. The quantitation of dsDNA is a very important step in many biological applications, particularly in standard molecular biology techniques. The dye shows minimal binding to single stranded DNA (ssDNA) and RNA, allowing specific quantitation of dsDNA.

 

  • Specificity: Highly specific to dsDNA, minimal binding to ssDNA, RNA, protein and interfering compounds.
  • Ease of Use: System includes all required reagents to quickly set up and quantitate dsDNA.
  • Instrument Compatibility: Easy to set up on the Quantus™, QuantiFluor™ and GloMax™-Multi Instruments. Also can be used on the Qubit™ instrument.

 

Order Info

Each QuantiFluor™ dsDNA System contains sufficient volume to perform 200 assays at 2mL or 2,000 assays at 200μL.

Specifications

For Use With (Application) Quantification of double stranded DNA using Fluorescent methods
Includes 25mL 20X TE Buffer (pH 7.5), 1mL 200X QuantiFluor™ dsDNA Dye, 100μg/mL 100μg lambda DNA standard
Quantity 1 mL

Patent pending. For Research Use Only. Not for Use in Diagnostic Procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.