Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega QuantiFluor™ RNA System

Catalog No. PRE3310 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
For Use With (Application):
Detects and quantifies small amounts of RNA in solution using a single-tube or plate reading fluorometer, downstream applications
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
PRE3310 Detects and quantifies small amounts of RNA in solution using a single-tube or plate reading fluorometer, downstream applications
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE3310 Supplier Promega Supplier No. E3310
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easily detects and quantifies small amounts of RNA in solution using a single-tube or plate reading fluorometer, supplied with QuantiFluor RNA Dye, Dilution Buffer and RNA standard

  • Contains fluorescent RNA-binding dye
  • For use in downstream applications
  • Quantitates low concentration pure RNA samples from 0.5ng/ml to 2,000ng/ml
  • Reproducible performance in downstream applications

Specifications

For Use With (Application) Detects and quantifies small amounts of RNA in solution using a single-tube or plate reading fluorometer, downstream applications
Includes 20X TE Buffer, 1 x 100μg RNA standard, 1 x 1mL QuantiFluor™ RNA dye
No. of Reactions 2000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.