Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell HT 96 gDNA Blood Isolation System

Catalog No. PRA2631 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 96 Preps
10 mL
4 x 96 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA2631 10 mL
PRA2671 4 x 96 Preps
PRA2670 1 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRA2631 Supplier Promega Supplier No. A2631
Add to Cart
Add to Cart

Includes

Binding Buffer, ReliaPrep Resin, Wash Buffer, 10mM EDTA (pH 8.0), 25mM Tris-HCl (pH 8.0), Proteinase K (PK) Solution, RNase A Solution, Cell Lysis Buffer (CLD)

The Maxwell HT 96 gDNA Blood Isolation System provides a simple and reliable method for the rapid isolation of gDNA in a multiwell format. The purified gDNA can be used directly in PCR assays, microarrays and next-generation sequencing applications.

  • Available in 0.5M or 10mM concentrations
  • 10mM concentration also provided as part of Maxwell(R) HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671)

Specifications

Content And Storage 15°C to 30°C
Format Multiwell
For Use With (Equipment) ReliaPrep™ 96 genomic DNA Miniprep HT system
For Use With (Application) Rapid isolation of genomic DNA
Includes Binding Buffer, ReliaPrep™ Resin, Wash Buffer, 10mL 10mM EDTA (pH 8), 25mM Tris HCl (pH 8), Proteinase K solution, RNase A solution, Cell Lysis Buffer
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.