Learn More
Description
Includes
Binding Buffer, ReliaPrep Resin, Wash Buffer, 10mM EDTA (pH 8.0), 25mM Tris-HCl (pH 8.0), Proteinase K (PK) Solution, RNase A Solution, Cell Lysis Buffer (CLD)
- High-Throughput DNA Extraction in 24-or 96-Well Format
- Robust, precipitation-free protocol, No chance of lost pellets.
- High yields of pure DNA from pristine and challenged or hemolysed samples.
- Pure gDNA ready for demanding applications; samples in solution; no resuspension required
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | Multiwell |
| Isolation Technology | Magnetic Bead |
| For Use With (Application) | Rapid isolation of genomic DNA |
| Includes | 5.5mL Proteinase K solution, 40mL Cell Lysis Buffer, 5.5mL ReliaPrep™ Resin, 50mL Binding Buffer, 85mL Wash Buffer, 60mL pH 8 25mM Tris HCl, 10mL pH 8 10mM EDTA, 1mL RNase A solution |
| No. of Reactions | 1 x 96 Preps |
| Product Type | gDNA Isolation Kit |
| Quantity | 1 x 96 Preps |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.