Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ReliaPrep™ RNA Miniprep System

Catalog No. PRZ6010 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 Preps
250 Preps
50 Preps
For Use With (Application):
Purify RNA from cultured cells
Purify RNA from tissues
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application)
PRZ6010 10 Preps Purify RNA from cultured cells
PRZ6012 250 Preps Purify RNA from cultured cells
PRZ6110 10 Preps Purify RNA from tissues
PRZ6112 250 Preps Purify RNA from tissues
PRZ6111 50 Preps Purify RNA from tissues
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PRZ6010 Supplier Promega Supplier No. Z6010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

ReliaPrep Minicolumns, Collection Tubes, 1-Thioglycerol, Column Wash Solution (CWE), Nuclease-Free Water, BL Buffer, Elution Tubes, RNA Wash Solution (RWA), Yellow Core Buffer, 0.09M MnCl2, DNase I (lyophilized)

The ReliaPrep RNA Miniprep Systems provide a fast and simple technique for preparation of intact total RNA from cultured cells or tissue in as little as 30 minutes from very small amounts of input material.

  • Efficiently Purify RNA from as Few as 100 Cells or 0.25mg Tissue
  • Simple, 30-minute protocol
  • Elutes in small volume, generating highly concentrated RNA
  • No phenol/chloroform or ß-mercaptoethanol required

Specifications

Content And Storage 15°C to 30°C
For Use With (Application) Purify RNA from cultured cells
Includes Collection tubes 2 x 10, ReliaPrep™ Minicolumns 1 x 10, 1-Thioglycerol 1 x 75μL, column wash solution (CWE) 1 x 1mL, Nuclease-free Water 1 x 1,250μL, BL Buffer 2 x 3.25mL, elution tubes 1 x 25, RNA was
Product Type RNA Kit
Quantity 10 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.