Learn More
Promega Nuclei Lysis Solution for Wizard™ Genomic DNA Purification Kits
Shop All Promega Corporation ProductsDescription
- Manual, Solution-Based DNA Extraction Method
- DNA from blood, animal and plant cells, bacteria or yeast
- 60-minute, scalable protocol
- Kit size based on number of isolations from whole blood, other sample types may vary
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| For Use With (Equipment) | Wizard™ genomic DNA purification kit |
| For Use With (Application) | Isolation of DNA from white Blood cells, tissue culture cells, animal tissue, plant tissue, yeast and gram positive and gram negative bacteria |
| Product Type | Cell Lysis Solution |
| Quantity | 1 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.