Learn More
Promega Wizard™ SV 96 Cell Resuspension Solution for Purification Systems
Catalog No. PRA7113 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Cell resuspension solution; use w/plasmid DNA purification systems; 500mL
Catalog No.
PRA7113 Cell resuspension solution; use w/plasmid DNA purification systems; 500mL
1 options
Catalog No. PRA7113 Supplier Promega Supplier No. A7113
Cell Resuspension Solution is a component of the Wizard™ MagneSil™, Wizard™ Plus and Wizard™ SV 96 and SV 9600 Plasmid DNA Purification Systems and the PureYield Plasmid Midiprep System.
  • Reagent Used in Plasmid DNA Purification Protocols
  • Cat.# A7112 is a component of the Wizard(R) Plus DNA Purification Systems
  • Cat.# A7115 is a component of the PureYield™ Plasmid Midiprep System
  • Cat.# A7113 and A7118 are components of the Wizard(R) SV 96 Plasmid DNA Purification System
  • Cat.# A7114 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System