Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Ribo m7G Cap Analog

Catalog No. p-132249 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
10 A254 U
25 A254 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-P1711 10 A254 U
PR-P1712 25 A254 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-P1711 Supplier Promega Supplier No. P1711
Only null left
Add to Cart
Add to Cart

This methylated ribonucleotide can be incorporated onto the 5'-end of in vitro transcripts and simulates the cap structure found on most eukaryotic mRNA molecules.

  • Enhances Translation Efficiency in Reticulocyte-Based Reactions
  • Protects RNA from intracellular digestion
  • Can be used with Riboprobe(R) or RiboMAX™ RNA production systems
  • Supplied at a concentration of 40mM

Specifications

Content And Storage -30°C to -10°C
Formulation Contains at least 13.7μL Ribo m7G cap analog in Nuclease-free Water
For Use With (Application) Enhances translation efficiency in many reticulocyte-based reactions and protects RNA from intracellular digestion
Quantity 10 A254 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.