Learn More
Promega Riboprobe™ Combination Systems
Catalog No. PRP1450 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
Riboprobe Combination System SP6/T7
Riboprobe Combination System T3/T7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PR-P1450 Riboprobe Combination System T3/T7
PR-P1460 Riboprobe Combination System SP6/T7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-P1450 Supplier Promega Supplier No. P1450
Add to Cart
Add to Cart
Includes
RNA polymerase(s); pGEM™ Express Positive Control Template; RQ1 RNase-free DNase; Recombinant RNasin™ Ribonuclease Inhibitor; rATP; rCTP; rGTP; rUTP; DTT; Transcription-optimized buffer, 5X; Nuclease-free water
The Riboprobe Systems are designed to prepare high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts.
  • Produce High-Specific-Activity RNA Probes or Defined RNA Transcripts
  • Choose from combination systems with T3/T7 or SP6/T7 RNA polymerases
  • SP6, T7 and T3 RNA Polymerases are extremely promoter-specific, allowing production of virtually homogeneous RNA using plasmid DNA as a template
  • Allows synthesis of RNA corresponding to the coding or noncoding strand of cloned DNA from a single plasmid construct
  • U.S. Patent 5,552,302; Australian Patent 646803 and other patents.
  • Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
Specifications
Content And Storage -30°C to -10°C
For Use With (Application) Generation of high-specific-activity RNA probes, generation of microgram quantities of RNA transcripts, generation of RNA processing substrates, generation of antisense RNA
Includes 500U T3 RNA Polymerase, 500U T7 RNA Polymerase, 5μg pGEM™ express positive control template, 500μL transcription optimized 5X Buffer, 2500U recombinant RNasin™ R
Product Type Riboprobe Combination System T3/T7
Quantity 1 System