Learn More
Description
- Designed for Sequencing Inserts Cloned into the pGEM(R), pALTER(R)-MAX and pCI-neo Vectors
- Designed to be annealed to single-stranded DNA or, after alkaline denaturation, to double-stranded DNA
- Purified by gel electrophoresis or HPLC
- Primer Sequence: 5′-d(TATTTAGGTGACACTATAG)-3′
- Supplied at a concentration of 10µg/ml
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.