Learn More
Promega RON Kinase Enzyme System
Catalog No. PRV8071 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8071 1/Ea.
PRV3921 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV8071 Supplier Promega Supplier No. V8071
Includes
  • Kinase: Recombinant Human RON, 10μg (amino acids 983-end), MW: ∽71kDa
  • Substrate: Axltide (KKSRGDYMTMQIG) derived from mouse Insulin receptor substrate 1 (amino acid 979 to 989)
  • Other: Reaction Buffer, DTT
Recombinant human RON (amino acids 983-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RON is a macrophage stimulating 1 receptor c-met-related tyrosine kinase.
  • Easily Screen and Profile RON Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Includes 1 x 10μg RON Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay
Content And Storage Components at different temps
Quantity 1/Ea.