Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega RSK2 Kinase Enzyme System

Catalog No. PRV3501 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3501 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV3501 Supplier Promega Supplier No. V3501
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human RSK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RSK2 is a member of the RSK (ribosomal S6 kinase) family that are growth factor-regulated serine-threonine kinases.

  • Easily Screen and Profile RSK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type Enzyme System
Includes Active Enzyme (G), Kinase Substrate, 0.1M DTT, 5x Reaction Buffer A
Content And Storage <-65°C
For Use With (Application) Mediates growth factor signaling, induces CREB serine-133 phosphorylation and activates gene expression
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.