Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega SGK1 Kinase Enzyme System

Catalog No. PRV9671 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ADP-Glo Kinase Assay and SGK1 Kinase System
SGK1 Kinase Enzyme System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV9671 1/Ea. ADP-Glo Kinase Assay and SGK1 Kinase System
PRV2911 10 μg SGK1 Kinase Enzyme System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9671 Supplier Promega Supplier No. V9671
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

SGK1 Kinase Assay:

  • Kinase: Recombinant Human SGK1, 10μg (amino acids 60 to end), MW: ≅73kDa
  • Substrate: Akt (PKB) substrate (CKRPRAASFAE)
  • Other: Reaction Buffer, DTT
ADP-Glo Kinase Assay: Reagent, buffer, substrate, Ultra Pure ATP (10mM), ADP (10mM)

Recombinant human SGK1 (amino acids 60-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SGK1 is a member of the serum- and glucocorticoid-induced protein kinase family.

  • Easily Screen and Profile SGK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type ADP-Glo Kinase Assay and SGK1 Kinase System
Molecular Weight (g/mol) 73
Content And Storage Components at different temps
For Use With (Application) For both primary screening as well as Kinase selectivity profiling, measure activity of recombinant Human SGK1
Quantity 1/Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.