Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Single-Step (KRX) Competent Cells

Catalog No. PRL3002 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRL3002 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRL3002 Supplier Promega Supplier No. L3002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Single Step (KRX) Competent Cells are designed for efficient transformation and tightly controlled protein expression with one strain.

  • Ready-to-Transform E. coli Strain with Tightly Controlled Protein Expression
  • Ready for protein expression two days after transformation
  • Dramatic control of expressed protein-coding regions with higher protein yields than BL21(DE3)-derived strains
  • Convenient blue/white screening for detecting recombinant clones

Specifications

Content And Storage <-65°C
Formulation Suspension in 10mM Tris HCl (pH 8), 100mM NaCl and 1mm EDTA
For Use With (Application) For Protein to Protein interaction studies which often involve generating numerous constructs, for cloning, screening and Protein expression
pH 7
Product Type Cell
Quantity 0.05 mL
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.