Learn More
Promega SRC Kinase Enzyme System
Catalog No. PRV9741 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9741 1/Ea.
PRV2921 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9741 Supplier Promega Supplier No. V9741
Includes
SRC Kinase Assay:
  • Kinase: Recombinant Human SRC, 10μg (full length), MW: ≅83kDa
  • Substrate: SRC substrate (KVEKIGEGTYGVVYK-amide) derived from human p34cdc2 (amino acid 6 to 20)
  • Other: Reaction Buffer, DTT, MnCl2
ADP-Glo Kinase Assay: Reagent, buffer, substrate, Ultra Pure ATP (10mM), ADP (10mM)
Recombinant full-length human SRC was expressed in E. coli cells using an N-terminal GST tag. SRC was originally identified as a transforming protein of the Rous sarcoma virus that had enzymatic ability to phosphorylate tyrosine in protein substrates.
  • Easily Screen and Profile SRC Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Includes 1 x 10μg SRC Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay (includes Reagent, Buffer, substrate, Ultra Pure ATP (10mM), ADP (10mM)
Content And Storage Components at different temps
Quantity 1/Ea.