Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega SRC Kinase Enzyme System

Catalog No. p-4672303 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2921 10 μg
PRV9741 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV2921 Supplier Promega Supplier No. V2921
Add to Cart
Add to Cart

Includes

SRC Kinase Assay:

  • Kinase: Recombinant Human SRC, 10μg (full length), MW: ≅83kDa
  • Substrate: SRC substrate (KVEKIGEGTYGVVYK-amide) derived from human p34cdc2 (amino acid 6 to 20)
  • Other: Reaction Buffer, DTT, MnCl2
ADP-Glo Kinase Assay: Reagent, buffer, substrate, Ultra Pure ATP (10mM), ADP (10mM)

Recombinant full-length human SRC was expressed in E. coli cells using an N-terminal GST tag. SRC was originally identified as a transforming protein of the Rous sarcoma virus that had enzymatic ability to phosphorylate tyrosine in protein substrates.

  • Easily Screen and Profile SRC Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) Increasing cell proliferation
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.