Learn More
Promega T7 RiboMAX™ Express Large Scale RNA Production System
Catalog No. PRP1320 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
50 x 20 μL Reactions
Catalog No. Quantity
PR-P1320 50 x 20 μL Reactions
1 options
Catalog No. PR-P1320 Supplier Promega Supplier No. P1320
Includes
pGEM™ Express Positive Control Template; RQ1 RNase-free DNase; Nuclease-free Water; T7 Express Enzyme Mix; Sodium Acetate, 3.0M (pH 5.2); RiboMAX Express T7 2X Buffer
The T7 RiboMAX Express Large Scale RNA Production System is an in vitro transcription system designed to produce milligram amounts of RNA in a short time.
  • Produces 5-8.5mg/mL RNA in 30 Minutes
  • Consistent production of milligram amounts of RNA
  • Minimal pipetting steps and setup time
  • Efficiently transcribes DNA templates of varying sizes. Works with transcripts as short as 21bp.
Specifications
Content And Storage -30°C to -10°C
For Use With (Application) Generation of large quantities of uncapped transcripts for in vitro or in vivo translation, in vitro synthesis of tRNA, rRNA, RNA virus genomes and ribozymes, production of substrates for RNA splicing, RNA secondary structure, antisense RNA and RNA:protein interaction studies; RNA for nonmammalian RNAi
Includes 100μL Enzyme Mix, T7 Express; 500μL RiboMAX Express T7 2X Buffer; 110U RQ1 RNase-free DNase; 2 × 5μg pGEM Express Positive Control Template; 1mL Sodium Acetate, 3.0M (pH 5.2); 1250μL Nuclease-free Water
Quantity 50 x 20 μL Reactions