Learn More
Promega Terminal Transferase Buffer Pack
Catalog No. PRM1893 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
3 x 500uL
Catalog No.
PRM1893 3 x 500uL
1 options
Catalog No. PRM1893 Supplier Promega Supplier No. M1893
TdT catalyzes the addition of mononucleotides to the terminal 3′-OH of a DNA initiator accompanied by the release of inorganic phosphate.
  • Adds Mononucleotides to the Terminal 3′-OH of ssDNA and Releases PPi
  • Adds mononucleotide tails to any type of 3′ end (3′ and 5′ overhangs or blunt ends) due to the presence of 1mM CoCl2 in the reaction buffer
  • Quality tested for labeling of apoptotic DNA ends using the procedure outlined in the DeadEnd™ Fluorometric TUNEL System Technical Bulletin #TB235 for each lot of enzyme
  • Supplied at a concentration of 30u/µL