Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega TNT™ Coupled Reticulocyte Lysate Systems

Catalog No. PRL4610 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
TnT SP6 Coupled Reticulocyte Lysate System
TnT T7 Coupled Reticulocyte Lysate System
No. of Reactions:
40
8 Reactions - Trial Size
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type No. of Reactions
PR-L4610 TnT T7 Coupled Reticulocyte Lysate System 40
PR-L4600 TnT SP6 Coupled Reticulocyte Lysate System 40
PR-L4611 TnT T7 Coupled Reticulocyte Lysate System 8 Reactions - Trial Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR-L4610 Supplier Promega Supplier No. L4610
Only null left
Add to Cart
Add to Cart

Includes

TNT Rabbit Reticulocyte Lysate, Appropriate TNT polymerase, Control DNA, TNT Reaction Buffer, Luciferase assay wells, Luciferase assay reagent, Amino acid mixtures

TNT Coupled Reticulocyte Lysate Systems are single-tube, coupled transcription/translation systems for eukaryotic cell-free protein expression.

  • A Single-Tube, Coupled Transcription/Translation System
  • Study protein:protein, protein:nucleic acid and protein expression interactions
  • Generate proteins in one hour versus days
  • You choose the promoter

  • U.S. Patents 5,324,637 and 5,492,817; Australian Patent 660329.
  • U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289.
  • Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.

Specifications

Content And Storage <-65°C
For Use With (Application) GST pull-downs, Gel shift assays, co-immunoprecipitation, characterization of Protein modifications, Protein-RNA interactions, Protein activity studies, confirmation of gene products, Protein truncation test (PTT)
No. of Reactions 40
Product Type TnT T7 Coupled Reticulocyte Lysate System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.