Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Thermal Printer Paper

Catalog No. PRE2851 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE2851 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE2851 Supplier Promega Supplier No. E2851
Add to Cart
Add to Cart

An ultra sensitive, compact laboratory instrument for measuring luminescent materials

  • For Use With TD-20/20 Instrument
  • Accessories only for discontinued Turner BioSystems Luminometer Model TD-20/20
  • Not for Turner BioSystems 20/20n Single-Tube Luminometer or Promega GloMax(R) 20/20 Luminometer
  • Power Supply now discontinued

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Turner BioSystems luminometer model TD-20/20
For Use With (Application) Measuring luminescent materials
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.