Learn More
Promega Ultra Pure Deoxynucleotide Triphosphates (dNTPs)
Catalog No. PRU1222 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
dCTP; 2 x 100umol; Concentration: 100mM
Catalog No.
PRU1222 dCTP; 2 x 100umol; Concentration: 100mM
1 options
This item is not returnable. View return policy
Catalog No. PRU1222 Supplier Promega Supplier No. U1222
This item is not returnable. View return policy
High-quality dNTPs. Provided at a concentration of 100mM in water at pH 7.5.
  • High-Quality Nucleotides for Your Workflow
  • Greater than 99% triphosphate content
  • 100mM in water at pH 7.5