Learn More
Promega Vac-Man™ 96 Vacuum Manifold
Catalog No. PRA2291 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
PR-A2291 1/Ea.
1 options
Catalog No. PR-A2291 Supplier Promega Supplier No. A2291
The Vac-Man 96 Vacuum Manifold is designed for manual use with the Promega SV 96 and SV 9600 Systems for isolating up to 96 samples simultaneously.
  • Eliminates the need to handle waste during nucleic acid purification
  • Adapts for use alone or with robotic nucleic acid purification platforms
  • Includes Vacuum Manifold Base, Manifold Collar and Manifold Bed
Specifications
Content And Storage 15°C to 30°C
Product Type Vac-Man 96 Vacuum Manifold
Quantity 1/Ea.