Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Vac-Man™ Vacuum Adapter

Catalog No. PRA1331 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-A1331 20 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-A1331 Supplier Promega Supplier No. A1331
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Required for use with the Wizard™ SV minicolumns for vacuum-based protocols.

  • Component of the Wizard Plus SV Minipreps DNA Purification Systems (Cat.# A1340 and A1470)
  • Enables use of vacuum-based plasmid miniprep protocol

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Silica Membrane Technology
For Use With (Equipment) Wizard™ Plus SV Minipreps start-up kit
For Use With (Application) Simple and reliable method for rapid isolation of plasmid DNA
Product Type Accessory
Quantity 20 Pack
Size 20 each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.