Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ViviRen™ In Vivo Renilla Luciferase Substrate

Catalog No. p-3578561 Shop All Promega Corporation Products
Click to view available options
Substrate Quantity:
0.37mg
3.7mg

ViviRen in vivo Renilla Luciferase Substrate is a uniquely engineered coelenterazine-based compound with protected oxidation sites. These modifications are designed to minimize substrate degradation and autoluminescence.

  • Engineered Coelenterazine-Based Substrate for Animal Imaging
  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings

Specifications

For Use With (Application) In-vivo bioluminescence imaging applications
Quantity 3.7mg
Substrate Type Renilla Luciferase Substrate
Storage Requirements −30 to −10°C
Substrate Quantity 3.7mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.