Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega VivoGlo™ Caspase-3/7 Substrate (Z-DEVD-Aminoluciferin, Sodium Salt)

Catalog No. PRP1782 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
50 mg
5 x 50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRP1782 5 x 50 mg
PRP1781 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRP1782 Supplier Promega Supplier No. P1782

VivoGlo™ Caspase-3/7 Substrate (Z-DEVD-Aminoluciferin, Sodium Salt) is a firefly luciferase prosubstrate containing the DEVD tetrapeptide sequence recognized by caspase-3 and -7.

  • Firefly Luciferase Prosubstrate Recognized and Cleaved by Caspase-3 and -7
  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings

Specifications

Molecular Weight (g/mol) 871.9
Content And Storage -30°C to -10°C
For Use With (Application) Eliminate potential interference in assays due to the presence of endotoxins, in-vivo bioluminescence imaging applications
Quantity 5 x 50 mg

  • The use of this product and derivatives thereof is strictly limited to that of a life sciences research reagent. All other use is strictly prohibited, including but not limited to any diagnostic, therapeutic, or commercial use. The use of this product for in vivo bioluminescent imaging may be covered by one or more patents controlled by Caliper Life Sciences, Inc. Those patents include U.S. Patents 5,650,135; 6,217,847; 6,923,951; 6,908,605; 6,890,515; and corresponding foreign patents and pending applications. The purchase or transfer of this product is not intended, either expressly or by implication, to grant any right or license to practice under the foregoing patents. Purchasers are advised to contact Caliper Life Sciences, Inc., 68 Elm St., Hopkinton, MA 01748 for information regarding a license to practice under such patents.
  • This product does not convey a license to use recombinant Coleoptera luciferase under U.S. Patents 5,583,024; 5,674,713 and 5,700,673. Promega sells licensed Coleoptera luciferase vectors, which may be used in conjunction with this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.