Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega VivoGlo™ In Vivo Imaging Substrates

VivoGlo™ Luciferin is the potassium salt of D-luciferin, the firefly luciferase substrate capable of generating light when a suitable model is used.

$540.76 - $1982.88

Specifications

For Use With (Application) In-vivo bioluminescence imaging applications
Substrate Type VivoGlo Luciferin
Storage Requirements −30 to −10°C
View More Specs

Products 3
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
PRP1041
View Documents
Promega
P1041
50mg
Each for $540.76
Only null left
Add to Cart
 
PRP1042
View Documents
Promega
P1042
250mg
Each for $1,071.92
Only null left
Add to Cart
 
PRP1043
View Documents
Promega
P1043
1g
Each for $1,982.88
Only null left
Add to Cart
 
Description

Description

  • Firefly Luciferase Substrate for Use in Animal Imaging Studies
  • High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
  • Available in multiple sizes to accommodate a variety of experimental settings
Specifications

Specifications

In-vivo bioluminescence imaging applications
VivoGlo Luciferin
−30 to −10°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.