Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ MagneSil™ Tfx™ System

Catalog No. PRA2380 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Wizard Magnesil Tfx System; 4 x 96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRA2380 Wizard Magnesil Tfx System; 4 x 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA2380 Supplier Promega Supplier No. A2380
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

MagneSil RED, endotoxin removal resin, MagneSil BLUE, 4/40 wash solution, cell resuspension solution (CRA), cell lysis solution (CLA), neutralization solution (NSA), nuclease-free water

The Wizard™ MagneSil™ Tfx System provides a simple method for the rapid isolation of transfection-quality plasmid DNA in a 96-well, high-throughput format.

  • Transfection-Quality Plasmid DNA in 96-Well Format
  • Improved transfection and protein expression results
  • Automated methods available

  • U.S. Patent 6,194,562; Australian Patent 740145; Canadian Patent 2,329,067 and other patents pending.
  • U.S. Patents 6,027,945; 6,368,800 and 6,673,631; Australian Patent 732756 and other patents and patents pending.
  • U.S. Patents 6,284,470 and 7,078,224; Australian Patent 778486 and other patents pending.

Specifications

Content And Storage 15°C to 30°C
Format 96-well Plate
Isolation Technology Magnetic Bead
For Use With (Application) Provides a simple and reliable method for the rapid isolation of transfection-quality plasmid DNA
Includes 150mL Cell resuspension solution, 150mL Cell Lysis solution, 150mL neutralization solution, 38mL Magnesil™ blue, 19mL endotoxin removal Resin, 30mL Magnesil™ red, 38mL wash solution, 2 x 25mL Nuclease-free Water, protoc
No. of Reactions 4 x 96 Preps
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.