Learn More
Promega Wizard™ PureFection Plasmid DNA Purification System1, 2
Catalog No. PRA2150 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
10 midipreps/2 maxipreps
125 midipreps/25 maxipreps
25 midipreps/5 maxipreps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-A2150 10 midipreps/2 maxipreps
PR-A2160 25 midipreps/5 maxipreps
PR-A2170 125 midipreps/25 maxipreps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR-A2150 Supplier Promega Supplier No. A2150
Employs a proprietary Endotoxin Removal Resin to produce purified plasmid DNA with low levels of endotoxin contamination
  • Utilizes MagneSil Paramagnetic Particles, a magnetic separation technology that allows rapid purification
  • Suitable for both midipreps (5 to 25mL of bacterial culture) and maxipreps (26 to 125mL of bacterial culture)
  • Produces DNA suitable for use in a variety of molecular biology applications, including transfection
  • Does not require phenol extractions or precipitations
  • Store at 22° to 25°C
Note: PR-A2160 and PR-A2170 do not contain the magnetic separation unit (PR-A2231); this unit must be purchased separately
  1. U.S. Patents 6,027,945; 6,368,800; and 6,673,631; Australian Patent 732756 and other patents and patents pending.
  2. U.S. Patent 6,194,562; Australian Patent 740145; Canadian Patent 2,329,067 and other patents pending.
Specifications
Quantity 10 midipreps/2 maxipreps