Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ SV 96 Binding Plates

Catalog No. PRA2278 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-A2278 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-A2278 Supplier Promega Supplier No. A2278
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Wizard™ SV 96 Binding Plates are used with several Wizard™ DNA purification systems and the SV 96 Total RNA Purification System.

  • Use with several Wizard(R) DNA purification systems or the SV 96 Total RNA Isolation System
  • Perform isolation manually or on a robotic platform
  • Use with the Vac-Man(R) 96 Vacuum Manifold

Specifications

Format Standard
Content And Storage 15°C to 30°C
Array 8 x 12
No. of Wells 96
For Use With (Application) Nucleic acid purification
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.