Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ SV Gel and PCR Clean-Up Start-Up Kit

Catalog No. PRA6750 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA6750 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA6750 Supplier Promega Supplier No. A6750
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • SV Gel and PCR Clean-Up System (SV Minicolumns, Collection Tubes, Membrane Wash Solution, Membrane Binding Solution, Nuclease-Free Water)
  • Welch™ Vacuum Pump for Continental Europe Electrical
  • Vac-Man™ Laboratory Vacuum Manifold (20-sample capacity)
  • Vacuum Adapters

The Wizard™ SV Gel and PCR Clean-Up System is designed to purify DNA fragments of 100bp-10kb from standard or low-melting agarose gels or to purify products directly from PCR or restriction digests.

  • Bundle of 500 DNA Purification Kits, Vacuum Manifold and Free Vacuum Pump
  • Purify 100bp to 10kb DNA from Gel Slices or PCR
  • Centrifuge or vacuum-based protocols

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Silica Spin Column
For Use With (Application) Extract and purify DNA fragments of 100bp to 10kb
Includes SV Gel and PCR clean-Up system, Welch™ vacuum pump for continental Europe electrical, Vac-Man™ laboratory vacuum manifold, vacuum adapters
Product Type DNA Purification Kit
Quantity Each
Voltage 110V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.